Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) carbonate basic, pure, 54-56% Cu, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Glass bottle
12069-69-1
CCuO3·CuO2H2
221.1
MFCD00010976
Copper(II)hydroxide carbonate
12069-69-1
CCuO3·CuO2H2
221.1
MFCD00010976
Copper(II)hydroxide carbonate
Linear Formula CuCO3·Cu(OH)2
Fieser 01,163
Merck Index 15, 2621
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids,ammonia,practically insoluble in alcohol
Formula Weight 221.1
Physical Form Fine Crystalline Powder
Percent Purity 54-56% Cu
Grade Pure
Density 4
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in acids, ammonia, practically insoluble in alcohol
Color Green
Melting Point 200.0°C
Quantity 25g
Chemical Name or Material Copper(II) carbonate basic
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position c

GHS Signal Word: Warning

EINECSNumber : 235-113-6

RTECSNumber : GL6910000

TSCA : TSCA