Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) carbonate dihydroxide, Cu 55% min

Catalog No. AA33305A3
Click to view available options
Quantity:
500 g
2 kg

CAS: 12069-69-1 | CH2Cu2O5

Copper(II) carbonate dihydroxide is used as pigments and in treatment for copper deficiency in ruminants. It is also used in pyrotechnics, in sweetening of petrol sour crude stock and animal and poultry feeds. It finds application in some types of make-up such as lipstick. In analytical chemistry, it is used as a reagent for analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
Molecular Formula CH2Cu2O5
MDL Number MFCD00010976
Synonym Copper(II) carbonate, basic, Copper(II) hydroxide carbonate

Specifications

Melting Point 200°C (decomposition)
Color Green
Physical Form Powder
Quantity 2 kg
Solubility Information Soluble in acid,ammonia,ammonium salt and alkali metal carbonate solution. Insoluble in water and alcohol.
Formula Weight 221.11
Density 4
Chemical Name or Material Copper(II) carbonate dihydroxide, Cu 55% min.
Hazard Category H302+H332-H319
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 235-113-6
RTECSNumber GL6910000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.