Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) chloride, anhydrous, 98% min., Alfa Aesar™

For Isomerization and Cracking Catalyst, Mordant in Dyeing and Printing Fabrics, in Electroplating Baths For Plating Cu On Al

Manufacturer:  Alfa Aesar 1245718

Quantity 50g

 View more versions of this product

Catalog No. AA1245718


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7447-39-4
98% min.
134.45
Copper(II) chloride, Anhydrous
Powder
CuCl2
MFCD00010972

Specifications

7447-39-4
98% min.
Yellow-brown powder
MFCD00010972
Odorless
50g
Powder
CuCl2
134.45
3.386g/cm3
Yellow-brown
Copper(II) chloride, Anhydrous

Safety and Handling

SDS & Safety Documents

H301-H314-H318-H400-H410; Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-210-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit