Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) chloride, 99%, extra pure, anhydrous

Catalog No. AC206530010
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 7447-39-4 | Cl2Cu | 134.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-39-4
Molecular Formula Cl2Cu
Molecular Weight (g/mol) 134.45
MDL Number MFCD00010972

Specifications

Boiling Point 993.0°C
Melting Point 498.0°C
Color Brown
Physical Form Fine Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5% min. (CuCl2)
Linear Formula CuCl2
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 10,106; 11,267; 13,152; 14,100
Merck Index 15, 2623
Synonym Cupric chloride
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 134.45
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Copper(II) chloride, anhydrous
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical
EINECSNumber 231-210-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.