Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) chloride dihydrate, 99%, extra pure, powder

Catalog No. p-222909
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Melting Point 598°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula CuCl2·2H2O
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 13,152
Merck Index 15, 2623
Solubility Information Solubility in water: 1150g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in acetone and ethylacetate, slightly soluble in ether
IUPAC Name dichlorocopper;dihydrate
Formula Weight 170.48
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Density 2.54
Chemical Name or Material Copper(II) chloride dihydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
RTECSNumber GL7030000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.