Learn More
Copper(II) chloride dihydrate, ACS, 99+%
Description
Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 100°C (decomposition) |
Physical Form | Granules |
Quantity | 500 g |
Assay Percent Range | >99% |
Solubility Information | Soluble in water,methanol and ethanol. |
Formula Weight | 170.48 (134.45anhydrous) |
Grade | ACS Reagent |
Sensitivity | Air sensitive; Hygroscopic |
Density | 2.51 |
Chemical Name or Material | Copper(II) Chloride Dihydrate |
Safety and Handling
Hazard Category | H302+H312-H315-H318 |
Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 231-210-2 |
RTECSNumber | GL7030000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.