Learn More
Copper(II) chloride, ultra dry, 99.995% (metals basis)
Description
Copper(II) chloride is used as a co-catalyst with palladium(II) chloride used in the Wacker process to prepare ethanol from ethane. It acts as an effective catalyst for the tetrahydropyranylation of alcohols. It is involved in the production of vinyl chloride and dichloroethane as well as in the chlorination of aromatic hydrocarbons. It acts as a deodorizing and desulfurizing agent in petroleum industry, mordant in textiles industry and in electrotyping baths for plating copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 993°C (decomposition) |
| Melting Point | 620°C |
| Color | Brown-Yellow |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.995% (Metal basis) |
| Solubility Information | Soluble in water,alcohol,acetone and hot sulfuric acid. |
| Formula Weight | 134.45 |
| Odor | Odorless |
| Sensitivity | Air sensitive; Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H315-H318 |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
| DOTInformation | DOT Class: 8, Packing Group: III |
| EINECSNumber | 231-210-2 |
| RTECSNumber | GL7000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.