Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) fluoride, 98%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
7789-19-7
CuF2
101.54
Cupric fluoride
7789-19-7
CuF2
101.54
Cupric fluoride
Assay Percent Range 97.5% min
Linear Formula CuF2
Merck Index 15,2628
Solubility Information Solubility in water: 47g/L (20°C). Other solubilities: hydrolyzes in hot water (forming oxyfluoride),soluble in acids and alcohol
Formula Weight 101.54
Physical Form Powder
Percent Purity 98%
Density 4.2300g/mL
Loss on Drying 0.5% max.
Packaging Plastic bottle
Solubility Solubility in water: 47g/L (20°C). Other solubilities: hydrolyzes in hot water (forming oxyfluoride), soluble in acids and alcohol
Specific Gravity 4.23
Color Blue-Gray to Gray
Melting Point 950.0°C
Quantity 25g
Chemical Name or Material Copper(II) fluoride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Contact with acids liberates very toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

EINECSNumber : 232-147-3