Learn More
Copper(II) hydroxide, tech. 94%, stab.
Description
Used as a raw material in chemical processing and catalyst manufacturing. Copper(II) hydroxide is used as ceramic colorant. In organic synthesis, it is used as a catalyst to prepare 1-((2-aminoethyl)amino)anthraquinone, 1-amino-4-((2-aminoethyl)amino)anthraquinone by reacting with 1-bromoanthraquinone or 1-amino-4-bromoanthraquinone respectively. It is involved in the synthesis of benzoic acid and octanoic acid. It is used as a precursor in the production of rayon, battery electrodes and other copper salts It is also used as a dye mordant, paper colorant, pigment and feed additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Blue |
| Physical Form | Powder |
| Quantity | 2 kg |
| Assay Percent Range | 94% |
| Solubility Information | Slightly soluble in waterSoluble in acids,ammonium hydroxide,dilute hydrochloric acid,concentrated alkali and potassium cyanide. Insoluble in water,ethanol,and acetone. |
| Formula Weight | 97.55 |
| Grade | Technical |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Blue powder |
| Show More |
Safety and Handling
| Hazard Category | H302-H318-H330 |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P301+P312-P304+P340-P305+P351+P338-P310-P330-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 243-815-9 |
| RTECSNumber | GL7600000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.