Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%

Catalog No. p-3256989
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg

For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths | CAS: 19004-19-4 | CuN2O6·2.5H2O

Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black and producing a burnished effect on ironCopper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4
Molecular Formula CuN2O6·2.5H2O
MDL Number MFCD00149671
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

Boiling Point 170°C (decomposition)
Melting Point ∽115°C
Physical Form Crystalline Lump
Quantity 50 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and alcoholSoluble in water,ammonia and ethanol.
Formula Weight 232.59 (187.55 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Density 2.32 g/cm3
Chemical Name or Material Copper(II) nitrate hemi(pentahydrate), trihydrate
Show More Show Less
Hazard Category H272-H314-H335
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1; Packaging Group: II
EINECSNumber 221-838-5
RTECSNumber GL7875000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.