Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) oxalate hemihydrate, 98%, Alfa Aesar™

Click to view available options
Quantity:
50g
250g
5893-66-3
C4H6Cu2O9
325.175
MFCD00150451
LIZLXPYGKGKSBP-UHFFFAOYSA-N
cupric oxalate,copper oxalate hemihydrate
122130000
copper;oxalic acid;hydrate
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Cu].[Cu]
5893-66-3
C4H6Cu2O9
325.175
MFCD00150451
LIZLXPYGKGKSBP-UHFFFAOYSA-N
cupric oxalate,copper oxalate hemihydrate
122130000
copper;oxalic acid;hydrate
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Cu].[Cu]
Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

Solubility
Soluble in NH{4}OH

Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
Quantity 250g
Assay Percent Range 98%
UN Number UN3288
Merck Index 14,2645
Solubility Information Soluble in NH4OH
Formula Weight 160.57 (151.51 Anhydrous)
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Copper(II) oxalate hemihydrate

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 212-411-4

TSCA : Yes

Recommended Storage : Ambient temperatures