Learn More
Copper(II) oxide, 97%
Description
As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactionsCopper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1326°C |
Color | Black |
Physical Form | Powder |
Quantity | 10 kg |
Assay Percent Range | 97% |
Solubility Information | Insoluble in water and alcohol. Soluble in dil. acids,NH4Cl and KCNPractically insoluble in water,and alcohol; soluble in dilute acids,alkali cyanides,and ammonium carbonate solution. |
Formula Weight | 79.55 |
Refractive Index | 2.63 |
Appearance | Black powder |
Density | 6.3 |
Show More |
Safety and Handling
Hazard Category | H400-H410-H302; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. |
DOTInformation | DOT Class: 9, Packaging Group: III |
EINECSNumber | 215-269-1 |
RTECSNumber | GL7900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.