Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) oxide, ACS, 99.0% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 3330730

Quantity 250g

 View more versions of this product

Catalog No. AA3330730


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1317-38-0
99.0% min.
79.55
Copper(II) oxide, For ACS analysis
Powder
CuO
MFCD00010979
ACS Grade

Specifications

1317-38-0
99.0% min.
Black powder
MFCD00010979
Odorless
250g
ACS Grade
Powder
CuO
79.55
6.315g/cm3
Black
Copper(II) oxide, For ACS analysis

Safety and Handling

SDS & Safety Documents

H400-H410-H302; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-269-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit