Learn More
Copper(II) oxide, nanopowder
Description
Copper(II) oxide nanopowder is used as a burning rate catalyst in rocket propellant, which enhances the the homogeneous propellant burning rate, lower pressure index. It can act as a catalyst used in superconducting materials, thermoelectric materials, sensing materials, glass and ceramics. It plays a vital role in photoconductive and photothermal applications. It is also used in ceramic resistors, magnetic storage media, gas sensors, near-infrared tilters, semiconductors, solar energy transformation and high-tech superconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1326°C |
| Color | Black |
| Physical Form | Powder |
| Quantity | 50 g |
| UN Number | UN3077 |
| Merck Index | 14,2646 |
| Solubility Information | Practically insoluble in water,and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution. |
| IUPAC Name | oxocopper |
| Formula Weight | 79.55 |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | GHS P Statement P264-P270-P301+P312-P330-P501a Wash thoroughly after handling. Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 215-269-1 |
| RTECSNumber | GL7900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.