Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) phthalocyanine

Catalog No. aa4365009
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. AA4365009

Supplier: Thermo Scientific Chemicals 04365009

Only null left
Add to Cart

CAS: 147-14-8 | C32H16CuN8

Copper(II) phthalocyanine are involved in the study of photosensitizer chemistry for uniform polymerization, luminescence chemistry and spectrophotometric analysis, organic synthesis and polymerization. This pigment is used in enamels, linoleum, inks, plastics, and rubber goods. Photoisomerizable phthalocyanines are used in rewritable CD or DVD printing. Other applications in organic solar cells, biosensitizers and display devices such as OLED , OTFT, Wearable Display, and e-paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym C.I. 74160; Pigment Blue 15

Specifications

Melting Point 350°C
Color Blue to Purple
Quantity 10 g
Beilstein 4121848
Merck Index 14,2520
Solubility Information It is insoluble in water.
Formula Weight 576.08
Physical Form Powder
Chemical Name or Material Copper (II) phthalocyanine
EINECSNumber 205-685-1
RTECSNumber GL8510000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.