Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) selenide, 99.5% (metals basis)

Catalog No. p-4781882
Click to view available options
Quantity:
5 g
25 g

For Use in Semiconductors, As Catalyst in Kjeldahl Digestion | CAS: 1317-41-5 | CuSe

In semiconductors, as catalyst in Kjeldahl digestion.Copper(II) selenide is involved in the preparation of nanocrystals, which shows significant photothermal heating with a photothermal transduction efficiency, thereby it is used for photothermal therapy applications. It is used as a semiconductor and ideal solar control coating as well as finds application in solar cell technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Form: 6mm and down pieces
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-41-5
Molecular Formula CuSe
MDL Number MFCD00016058

Specifications

Melting Point 550°C (decomposition)
Color Black
Physical Form Pieces
Quantity 5 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Soluble in HCl with H2Se evolution,in H2SO4 with SO2 evolution. Oxidized to CuSeO3 by HNO3Soluble in hydrochloric acid,sulfuric acid. Insoluble in water.
Formula Weight 142.5
Density 5.99 g/cm3
Chemical Name or Material Copper(II) selenide
Hazard Category H301+H331-H373
Hazard Statement When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-272-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.