Copper(II) selenide, Black pieces, 99.5% (Metals basis), Alfa Aesar™

For Use in Semiconductors, As Catalyst in Kjeldahl Digestion

Manufacturer: Alfa Aesar 1790114

Quantity 25g

 View more versions of this product

Catalog No. AA1790114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-41-5
    Physical Form Pieces
    Assay Percent Range 99.5% (Metals basis)
    Molecular Formula CuSe
    Formula Weight 142.5
    MDL Number MFCD00016058
    Chemical Name or Material Copper(II) selenide

    Specifications

    Physical Form Pieces
    Formula Weight 142.5
    Quantity 25g
    Color Black
    DOT Information DOT Class: 9, Packaging Group: III
    EINECS Number 215-272-8
    Assay Percent Range 99.5% (Metals basis)
    Molecular Formula CuSe
    MDL Number MFCD00016058
    CAS 1317-41-5
    Health Hazard 1 H301-H331-H373-H400-H410; Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Health Hazard 2 When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
    Chemical Name or Material Copper(II) selenide

    When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

    H301-H331-H373-H400-H410; Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit