Learn More
Copper(II) sulfate pentahydrate, 99%
Description
Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.
Solubility
Soluble in water and methanol. Insoluble in alcohol.
Notes
Incompatible with steel, finely powdered metals, hydroxylamine, magnesium, hydrazine, nitromethane, alkalies, phosphates and acetylene gas
Specifications
Specifications
| pH | 4 |
| Quantity | 5000 g |
| UN Number | UN3288 |
| Merck Index | 14,2653 |
| Solubility Information | Soluble in water and methanol. Insoluble in alcohol. |
| Formula Weight | 249.68 (159.60 Anhydrous) |
| Percent Purity | 99% |
| Odor | Odorless |
| Density | 2.27 |
| Chemical Name or Material | Copper (II) sulfate pentahydrate |
Safety and Handling
| Hazard Category | H302-H315-H319 |
| Hazard Statement | GHS H Statement H301-H315-H319 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 231-847-6 |
| RTECSNumber | GL8900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.