Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) sulfate pentahydrate, Puriss., suitable for, meets analytical spec. of BP, FCC, Ph. Eur., USP, Honeywell Fluka™

Catalog No. 6000464 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 1 kg

Puriss., suitable for, Meets analytical specification of BP, FCC, Ph. Eur., USP

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Quantity 2.5 kg
Linear Formula CuSO4 · 5H2O
UN Number 3077
IUPAC Name copper;sulfate;pentahydrate
Formula Weight 249.69g/mol
Grade puriss.
Vapor Pressure 7.3mmHg (25°C)
Chemical Name or Material Copper(Ii) Sulfate Pentahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H315-H319-H410
EINECSNumber 231-847-6
RTECSNumber GL8900000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.