Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) tetrafluoroborate hexahydrate, 98%

Catalog No. p-3256991
Click to view available options
Quantity:
100 g
500 g

CAS: 72259-10-0 | B2CuF8H12O6 | 345.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72259-10-0
Molecular Formula B2CuF8H12O6
Molecular Weight (g/mol) 345.24
MDL Number MFCD05863735
InChI Key FBYIXFBIARQMJJ-UHFFFAOYSA-N
Synonym copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci
PubChem CID 15774553
IUPAC Name copper(2+) bis(tetrafluoroboranuide) hexahydrate
SMILES O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Blue
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 998%
Solubility Information Soluble in water.
Formula Weight 345.25 (237.15 Anhydrous)
Sensitivity Hygroscopic
Appearance Blue crystals
Chemical Name or Material Copper(II) tetrafluoroborate hexahydrate, hexahydrate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 253-959-4
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.