Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) trifluoroacetate hydrate

Catalog No. p-4407446
Click to view available options
Quantity:
10 g
50 g

CAS: 123333-88-0 | C4CuF6O4 | 289.58 g/mol

As Heterogeneous Catalysts. It is suitable for use as an electrocatalyst in gas diffusion electrodes. As Electrochemical gas sensors. As organic intermediate, as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-88-0
Molecular Formula C4CuF6O4
Molecular Weight (g/mol) 289.58
MDL Number MFCD00013200
InChI Key JIDMEYQIXXJQCC-UHFFFAOYSA-L
Synonym copper ii trifluoroacetate hydrate, 289.58 anhydrous, 2c2hf3o2.cu.xh2o, 2,2,2-trifluoroacetyl oxy cuprio 2,2,2-trifluoroacetate hydrate
PubChem CID 71311159
SMILES [Cu++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Physical Form Crystalline
Quantity 10 g
IUPAC Name copper;2,2,2-trifluoroacetic acid;hydrate
Formula Weight 289.57 (Anhydrous)
Chemical Name or Material Copper(II) trifluoroacetate hydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.