Copper iodate, anhydrous, 95%, Alfa Aesar™

Manufacturer: Alfa Aesar 8945904

Quantity 2g

 View more versions of this product

Catalog No. AA8945904

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13454-89-2
    Physical Form Powder
    Assay Percent Range 95%
    Molecular Formula Cu(IO3)2
    Formula Weight 413.35
    MDL Number MFCD00016055
    Chemical Name or Material Copper iodate, anhydrous

    Specifications

    Formula Weight 413.35
    Quantity 2g
    Color Green
    DOT Information DOT Class: 5.1, Packing Group: II
    Physical Form Powder
    Assay Percent Range 95%
    Molecular Formula Cu(IO3)2
    Appearance Green powder
    MDL Number MFCD00016055
    CAS 13454-89-2
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Copper iodate, anhydrous

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit