Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. p-218107 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle

Catalog No. SC194-100

For atomic absorption. (1mL = 1mg Cu) Solute: Copper Nitrate Solvent: Dilute Nitric Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4, 7697-37-2, 7732-18-5
MDL Number MFCD00010965

Specifications

Packaging Poly Bottle
Vapor Pressure 14mmHg at 20°C
Physical Form Liquid
Concentration 990 to 1010 ppm
Color Blue
Boiling Point 100°C
Quantity 100 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Copper, Reference Standard Solution, 1000ppm ±1%
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.