Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate Solution, 1.4% in 0.6% Sulfuric Acid, Reagents

Catalog No. 01335563
Change view
Click to view available options
:
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-563 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-563 Supplier Reagents Holdings Llc Supplier No. CC256200500A
Only null left
Add to Cart
Add to Cart

Copper Sulfate Solution, 1.4% in 0.6% Sulfuric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Copper Sulfate Solution, 1.4% in 0.6% Sulfuric Acid
Boiling Point 100°C
CAS 7732-18-5, 7758-99-8
Color Blue
pH <1
Physical Form Liquid
Quantity 500 mL
Molecular Formula CuSO4/H2O/H2SO4
Synonym Copper(II) Sulfate
Percent Purity 1.4%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.01
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.