Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper Sulfate Solution, 2% w/v, AquaPhoenix Scientific™

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
CAS | 7758-99-8, 7732-18-5 |
Quantity | 500 mL |
Concentration or Composition (by Analyte or Components) | 2% w/v |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction