Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate Solution for Aluminum Can Coating Test, Reagents

Catalog No. 01335667
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Poly Bottle
Catalog No. Quantity Packaging
01-335-667 4 L Glass Bottle
01-335-668 4 L Poly Bottle
2 options

Catalog No. 01-335-667

Supplier: Reagents Holdings Llc CC1021004C

Only null left
Add to Cart
Add to Cart

Copper Sulfate Solution for Aluminum Can Coating Test, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Copper Sulfate Solution for Aluminum Can Coating Test
Boiling Point 102°C
Color Blue/Green
CAS 7732-18-5, 7758-99-8
pH 1.98 to 2.16
Physical Form Liquid
Quantity 4 L
Molecular Formula CuSO4/H2O
Synonym Copper(II) Sulfate
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.