Copper Wire, 1.0mm (0.04 in.) dia, annealed, 99.9% (Metals basis), Alfa Aesar™

Form: Approx.7.02g/m

Manufacturer: Alfa Aesar 10782G9

Quantity 25m

 View more versions of this product

Catalog No. AA10782G9

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    Physical Form Wire, 1.0mm (0.04 in.) dia.
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00010965
    Chemical Name or Material Copper, Annealed

    Specifications

    Quantity 25m
    Color Copper colored
    Density 8.94g/cm3
    Physical Form Wire, 1.0mm (0.04 in.) dia.
    MDL Number MFCD00010965
    CAS 7440-50-8
    Assay Percent Range 99.9% (Metals basis)
    Odor Odorless
    Appearance Copper colored wire
    Chemical Name or Material Copper, Annealed

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit