Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper wire, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Click to view available options
Quantity:
10m
2m
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]

Form: approx. 2.87g/m

Quantity 2m
Appearance Copper colored wire
Density 8.94g/cm3
Assay Percent Range 99.999% (Metals basis)
Odor Odorless
Physical Form Wire, ≈2.87g/m
Vapor Pressure 0 hPa
Color Undesignated
Chemical Name or Material Copper

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures