Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104454 Supplier Thermo Scientific Supplier No. PA5104454
Add to Cart
Add to Cart

COPS3 Polyclonal Antibody

COPS3 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen COPS3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cops3
Gene Accession No. O88543, Q68FW9, Q9UNS2
Gene Alias COP9 (constitutive photomorphogenic) homolog, subunit 3; COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 3; COP9 complex S3; COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 signalosome complex subunit 3; COP9 signalosome subunit 3; COPS 3; COPS3; Csn3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3
Gene Symbols Cops3
Host Species Rabbit
Immunogen A synthesized peptide derived from human COPS3(Accession Q9UNS2), corresponding to amino acid residues E388-S423.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26572, 287367, 8533
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.