Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596546 Supplier Invitrogen™ Supplier No. PA596546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLKNFERALY FYEQAITTPA MAVSHIMLES YKKYILVSLI LLGKVQQLPK YTSQIVGRFI KPLSNAYHEL AQVYSTNNPS ELRNLVNKHS ETFTRDNNMG LVKQCLSSLY KKNIQRLTKT FLTLSLQDMA SRVQLSGPQE AEKYVLHMIE DGEIFASINQ KDGMVSFHDN PEKYNNPAML HNIDQEMLKC IELDERLKAM DQEITVNPQF VQKSMGSQED DSGNKPSSYS; Positive Samples: SW480, HeLa, 293T, Mouse heart, Rat heart, Rat brain; Cellular Location: Cytoplasm, Nucleus.

The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPS3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cops3
Gene Accession No. O88543, Q68FW9, Q9UNS2
Gene Alias COP9 (constitutive photomorphogenic) homolog, subunit 3; COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 3; COP9 complex S3; COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 signalosome complex subunit 3; COP9 signalosome subunit 3; COPS 3; COPS3; Csn3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3
Gene Symbols Cops3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 194-423 of human COPS3 (NP_0036442).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26572, 287367, 8533
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.