Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531125 Supplier Invitrogen™ Supplier No. PA531125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (94%), Xenopus laevis (96%), Pig (100%), Chicken (99%), Rhesus Monkey (100%), Chimpanzee (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes one of eight subunits composing COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPS4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene COPS4
Gene Accession No. Q9BT78
Gene Alias AW208976; COP9 (constitutive photomorphogenic) homolog, subunit 4; COP9 (constitutive photomorphogenic) homolog, subunit 4 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 4; COP9 complex S4; COP9 constitutive photomorphogenic homolog subunit 4; COP9 constitutive photomorphogenic-like protein subunit 4; COP9 signalosome complex subunit 4; COP9 signalosome subunit 4; COPS4; CSN4; D5Ertd774e; JAB1-containing signalosome subunit 4; SGN4; signalosome subunit 4; testis tissue sperm-binding protein Li 42a
Gene Symbols COPS4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human COPS4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.