Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS6 Antibody (OTI4E7), Alexa Fluor™ 647, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271648AF647

Catalog No. N271648A647


Only null left
Add to Cart

Description

Description

COPS6 Monoclonal specifically detects COPS6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

COPS6
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
COPS6
Recombinant protein fragment corresponding to amino acids 58-327 of human COPS6 (NP_006824) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI4E7
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
BMP2B, BMP2B1, BMP4, COP9 constitutive photomorphogenic homolog subunit 6 (Arabidopsis), COP9 signalosome complex subunit 6, CSN6COP9 subunit 6 (MOV34 homolog, 34 kD), H_NH0506M12.12, hVIP, JAB1-containing signalosome subunit 6, MOFC11, MOV34 homolog, MOV34 homolog, 34 kD, MOV34-34KD, SGN6, Signalosome subunit 6, Vpr-interacting protein
Mouse
Affinity Purified
DNA replication Transcription Translation and Splicing
10980.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.