Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577120 Supplier Invitrogen™ Supplier No. PA577120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The protein encoded by this gene is one of the eight subunits of COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. This protein belongs to translation initiation factor 3 superfamily. It is involved in the regulation of cell cycle and likely to be a cellular cofactor for HIV-1 accessory gene product Vpr.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPS6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene COPS6
Gene Accession No. O88545, Q7L5N1
Gene Alias COP9 (constitutive photomorphogenic) homolog, subunit 6; COP9 (constitutive photomorphogenic) homolog, subunit 6 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 6; COP9 complex S6; COP9 constitutive photomorphogenic homolog subunit 6; COP9 signalosome complex subunit 6; COP9 signalosome complex subunit 6-like protein; COP9 signalosome subunit 6; cops6; CSN6; H_NH0506M12.12; hVIP; hypothetical protein LOC550465; JAB1-containing signalosome subunit 6; MOV34 homolog; MOV34 homolog, 34 kD; MOV34-34KD; Sgn3; SGN6; signalosome subunit 6; VIP/MOV34; Vpr-interacting protein; zgc:112075
Gene Symbols COPS6
Host Species Rabbit
Immunogen Recombinant full length Human COPS6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10980, 26893, 304343
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.