Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS7A Antibody, Novus Biologicals™
SDP

Catalog No. NBP153118 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP153118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP153118 Supplier Novus Biologicals Supplier No. NBP153118
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COPS7A Polyclonal specifically detects COPS7A in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen COPS7A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UBW8
Gene Alias COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 7A, COP9 complex subunit 7a, COP9 constitutive photomorphogenic homolog subunit 7A (Arabidopsis), COP9 signalosome complex subunit 7a, CSN7AMGC110877, Dermal papilla-derived protein 10, DERP10, JAB1-containing signalosome subunit 7a, SGN7a, Signalosome subunit 7a
Gene Symbols COPS7A
Host Species Rabbit
Immunogen Synthetic peptides corresponding to COPS7A(COP9 constitutive photomorphogenic homolog subunit 7A (Arabidopsis)) The peptide sequence was selected from the middle region of COPS7A. Peptide sequence NLKKTIKVTTAAAAAATSQDPEQHLTELREPAPGTNQRQPSKKASKGKGL The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 30 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50813
Test Specificity This product is specific to Subunit or Isoform: 7a.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.