Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPS7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121179 Supplier Invitrogen™ Supplier No. PA5121179
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, HepG2, NCI-H460, A-549, Mouse brain, Mouse heart, Mouse kidney, Rat heart, Rat kidney. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MSAEVKVTGQ NQEQFLLLAK SAKGAALATL IHQVLEAPGV YVFGELLDMP NVRELAESDF ASTFRLLTVF AYGTYADYLA EARNLPPLTE AQKNKLRHLS VVTLAAKVKC IPYAVLLEAL ALRNVRQLED LVIEAVYADV LRGSLDQRNQ RLEVDYSIGR DIQRQDLSAI ARTLQEWCVG CEVVLSGIEE QVSRANQHKE QQLGLKQQIE SEVANLKKTI KVTTAAAAAA TSQDPEQHLT ELREPAPGTN QRQPSKKASK GKGLRGSAKI WSKSN.

The COP9 signalosome (CSN) complex is involved in several different developmental and cellular processes. The complex is made up of several widely expressed proteins: CSN1 (COPS1), CSN2 (COPS2), CSN3 (COPS3), CSN4 (COPS4), CSN5 (COPS5), CSN6 (COP6), CSN7a (COPS7, COPS7a) or CSN7b (COP7b) and CSN8 (COP8). The CSN complex acts as a regulator for the ubiquitin conjugation pathway by mediating the deneddylation of the SCF-type E3 ligase complexes, which leads to a decrease in ubiquitin ligase activity of SCF-complexes. It is also involved in the phosphorylation of p53, c-Jun, I B-alpha and IRF-8, as well as CSN-dependent phosphorylation of p53, and c-Jun protects and promotes degradation by the Ubl system. CSN7 is phosphorylated by CK2 and is composed of two subunits; a and b. CSN7a contains a PCI (Proteasome CSN9 initiation factor 3) region, as well as a coiled-coil region and is predicted to interact with CSN2, CSN3, CSN4, CSN5, CSN6, CSN8, and GPS1. CSN7b contains only a PCI region and is predicted to interact with INT6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPS7A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COPS7A
Gene Accession No. Q9CZ04, Q9UBW8
Gene Alias COP9 (constitutive photomorphogenic) homolog, subunit 7a; COP9 (constitutive photomorphogenic) homolog, subunit 7a (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 7a; COP9 complex S7a; COP9 complex subunit 7a; COP9 constitutive photomorphogenic homolog subunit 7A; COP9 signalosome complex subunit 7a; COP9 signalosome subunit 7A; COPS7A; CSN7; CSN7A; D6Ertd35e; dermal papilla-derived protein 10; DERP10; JAB1-containing signalosome subunit 7a; SGN7a; signalosome subunit 7a
Gene Symbols COPS7A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human COPS7A (NP_057403.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26894, 312710, 50813
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.