Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COPS8 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176373
Description
COPS8 Polyclonal specifically detects COPS8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
COPS8 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
COP9, COP9 constitutive photomorphogenic homolog subunit 8 (Arabidopsis), COP9 homolog, COP9 signalosome complex subunit 8, CSN8JAB1-containing signalosome subunit 8, hCOP9, MGC1297, MMMEP, SGN8MGC43256, Signalosome subunit 8 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 2 μg/mL, ELISA 1:100-1:2000 | |
NP_006701 | |
COPS8 | |
Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human CSN8. The immunogen is located within the last 50 amino acids of CSN8. Amino Acid Squence: PLSEPAPVPPIPNEQ | |
0.1 mg | |
Lipid and Metabolism | |
10920 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction