Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557682 Supplier Invitrogen™ Supplier No. PA557682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGLPVEEAVK GILEQGWQAD STTRMVLPRK PVAGALDVSF NKFIPLSEPA P Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.

COPS8 is a COP9 signalosome (CSN), which is an evolutionarily conserved protein complex of the eight subunits that interacts with deubiquitinating enzymes and protein kinases and is highly homologous to the lid sub-complex of 26S proteasome. The CSN complex is an essential regulator of the ubiquitin conjugation pathway by mediating the deneddylation of the SCF-type E3 ligase complexes, which leads to a decrease in ubiquitin ligase activity of SCF-complexes such as SCF, CSA or DDB2. It is also involved in phosphorylation of p53, c-jun/JUN, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN8 encodes the smallest and the least conserved but first identified subunit of CSN. Recent studies show CSN8 is essential for Drosophila development and is essential for peripheral T cell homeostasis and antigen receptor-induced entry into the cell cycle from quiescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPS8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cops8
Gene Accession No. Q99627
Gene Alias 9430009J09Rik; AA408242; COP9; COP9 (constitutive photomorphogenic) homolog, subunit 8; COP9 (constitutive photomorphogenic) homolog, subunit 8 (Arabidopsis thaliana); COP9 constitutive photomorphogenic homolog subunit 8; COP9 homolog; COP9 signalosome complex subunit 8; COP9 signalosome subunit 8; COPS8; Csn8; hCOP9; JAB1-containing signalosome subunit 8; SGN8; signalosome subunit 8
Gene Symbols Cops8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COPS8. Recombinant protein control fragment (Product #RP-96235).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.