Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581598
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581598 Supplier Invitrogen™ Supplier No. PA581598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat COPZ1/Zeta-1 COP.

This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPZ1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene COPZ1
Gene Accession No. P61923, P61924
Gene Alias 5930435A22Rik; AA407760; CGI-120; CHUNP6876; coatomer; coatomer protein complex subunit zeta 1; coatomer protein complex, subunit zeta 1; coatomer subunit zeta-1; coatomer subunit zeta-1-like protein; COPI coat complex subunit zeta 1; COPZ; COPZ1; D4Ertd360e; HSPC181; nonclathrin coat protein zeta1-COP; Zeta-1 COP; Zeta-1-coat protein; zeta1-cop; zeta-COP
Gene Symbols COPZ1
Host Species Rabbit
Immunogen Recombinant Rat COPZ1/Zeta-1 COP Protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22818, 315345, 56447
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.