Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597605
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597605 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597605 Supplier Invitrogen™ Supplier No. PA597605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COPZ1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPZ1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene COPZ1
Gene Accession No. P61923
Gene Alias 5930435A22Rik; AA407760; CGI-120; CHUNP6876; coatomer; coatomer protein complex subunit zeta 1; coatomer protein complex, subunit zeta 1; coatomer subunit zeta-1; coatomer subunit zeta-1-like protein; COPI coat complex subunit zeta 1; COPZ; COPZ1; D4Ertd360e; HSPC181; nonclathrin coat protein zeta1-COP; Zeta-1 COP; Zeta-1-coat protein; zeta1-cop; zeta-COP
Gene Symbols COPZ1
Host Species Rabbit
Immunogen Recombinant Human Coatomer subunit zeta-1 protein (1-177AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22818
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.