Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COQ10A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567027
Description
Immunogen sequence: YEVVSNVQEYR EFVPWCKKSL VVSSRKGHLK AQLEV Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
Required for the function of coenzyme Q in the respiratory chain. May serve as a chaperone or may be involved in the transport of Q6 from its site of synthesis to the catalytic sites of the respiratory complexes (Probable).
Specifications
COQ10A | |
Polyclonal | |
Unconjugated | |
COQ10A | |
coenzyme Q10 homolog A; coenzyme Q10 homolog A (S. cerevisiae); coenzyme Q10 homolog A (yeast); coenzyme Q10A; Coenzyme Q-binding protein COQ10 homolog A, mitochondrial; COQ10A; Gm1; RGD1562447; UNQ6192/PRO20219 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93058 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MF6 | |
COQ10A | |
Recombinant Human COQ10A. Recombinant protein control fragment (Product #RP-107666). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction