Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COQ10B Polyclonal Antibody, Bioss
SDP

Catalog No. 502281660
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1660 Supplier Bioss Supplier No. BS11656R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COQ10B Polyclonal antibody specifically detects COQ10B in Human, Mouse samples. It is validated for ELISA, Western Blot

Specifications

Antigen COQ10B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene COQ10B
Gene Accession No. Q3THF9, Q9H8M1
Gene Alias 1500041J02Rik; 1700030I21Rik; 9530077A17Rik; AV002237; AW742641; coenzyme Q10 homolog B; coenzyme Q10 homolog B (S. cerevisiae); coenzyme Q10B; coenzyme Q-binding protein COQ10 homolog B, mitochondrial; COQ10B; RGD1359509
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human COQ10B
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67876, 80219
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.