Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COQ2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305834IR
Description
COQ2 Polyclonal specifically detects COQ2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COQ2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
4-hydroxybenzoate polyprenyltransferase, coenzyme Q2 homolog, prenyltransferase (yeast), COQ2 homolog, EC 2.5.1, EC 2.5.1.39, FLJ13014, hCOQ2, mitochondrial, Para-hydroxybenzoate--polyprenyltransferase, para-hydroxybenzoate-polyprenyltransferase, mitochondrial, PHB:polyprenyltransferase | |
Produced in rabbits immunized with E. coli-derived Human COQ2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
27235 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction