Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COQ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107103 Supplier Invitrogen™ Supplier No. PA5107103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Dog: 93%; Goat: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 93%; Rat: 86%; Yeast: 91%; Zebrafish: 86%.

Catalyzes the prenylation of para-hydroxybenzoate (PHB) with an all-trans polyprenyl group. Mediates the second step in the final reaction sequence of coenzyme Q (CoQ) biosynthesis, which is the condensation of the polyisoprenoid side chain with PHB, generating the first membrane-bound Q intermediate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COQ2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene COQ2
Gene Accession No. Q96H96
Gene Alias 4-HB polyprenyltransferase; 4-hydroxybenzoate decaprenyltransferase; 4-hydroxybenzoate polyprenyltransferase, mitochondrial; CL640; coenzyme Q2 4-hydroxybenzoate polyprenyltransferase; coenzyme Q2 homolog, prenyltransferase; coenzyme Q2, polyprenyltransferase; COQ10D1; COQ2; MSA1; para-hydroxybenzoate-polyprenyltransferase, mitochondrial; PHB:polyprenyltransferase; PHB:PPT
Gene Symbols COQ2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human COQ2 (aa 238-288).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27235
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.