Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COQ3 Polyclonal Antibody
COQ3 Polyclonal Antibody
Supplier: Thermo Scientific PA556698
Description
COQ3 Polyclonal Antibody for IHC

Specifications
COQ3 | |
Polyclonal | |
Unconjugated | |
COQ3 | |
2-polyprenyl-6-hydroxyphenol methylase; 2-polyprenyl-6-hydroxyphenyl methylase; 3,4-dihydroxy-5-hexaprenylbenzoate methyltransferase; 3-demethylubiquinol 3-O-methyltransferase; 3-demethylubiquinone-10 3-methyltransferase; 3-demethylubiquinone-9 3-methyltransferase; 4732433J24; bA9819.1; C77934; Coenzyme Q (ubiquinone); coenzyme Q3 homolog methyltransferase; coenzyme Q3 homolog, methyltransferase; coenzyme Q3 homolog, methyltransferase (yeast); coenzyme Q3 methyltransferase; coenzyme Q3, methyltransferase; Coq3; DHHB methyltransferase; DHHBMT; DHHB-MT; DHHBMTASE; DHHB-MTase; dihydroxyhexaprenylbenzoate methyltransferase; hexaprenyldihydroxybenzoate methyltransferase, mitochondrial; methyltransferase COQ3; polyprenyldihydroxybenzoate methyltransferase; Ubiquinone biosynthesis O-methyltransferase, mitochondrial; UG0215E05 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51805 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZJ6 | |
COQ3 | |
Recombinant protein corresponding to Human COQ3. Recombinant protein control fragment (Product #RP-94829). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction