Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COQ3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556699 Supplier Invitrogen™ Supplier No. PA556699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WEKFVSPETL ESILESNGLS VQTVVGMLYN PFSGYWHWSE NTSLNYAAHA VKSRVQEHPA SAEFVLKGET EELQANACTN PAVHEKLKK Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 57%.

Ubiquinone, also known as coenzyme Q, or Q, is a critical component of the electron transport pathways of both eukaryotes and prokaryotes. This lipid consists of a hydrophobic isoprenoid tail and a quinone head group. The tail varies in length depending on the organism, but its purpose is to anchor coenzyme Q to the membrane. The quinone head group is responsible for the activity of coenzyme Q in the respiratory chain. The S. cerevisiae COQ3 gene encodes an O-methyltransferase required for 2 steps in the biosynthetic pathway of coenzyme Q. This enzyme methylates an early coenzyme Q intermediate, 3,4-dihydroxy-5-polyprenylbenzoic acid, as well as the final intermediate in the pathway, converting demethyl-ubiquinone to coenzyme Q. The COQ3 gene product is also capable of methylating the distinct prokaryotic early intermediate 2-hydroxy-6-polyprenyl phenol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COQ3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COQ3
Gene Accession No. Q9NZJ6
Gene Alias 2-polyprenyl-6-hydroxyphenol methylase; 2-polyprenyl-6-hydroxyphenyl methylase; 3,4-dihydroxy-5-hexaprenylbenzoate methyltransferase; 3-demethylubiquinol 3-O-methyltransferase; 3-demethylubiquinone-10 3-methyltransferase; 3-demethylubiquinone-9 3-methyltransferase; 4732433J24; bA9819.1; C77934; Coenzyme Q (ubiquinone); coenzyme Q3 homolog methyltransferase; coenzyme Q3 homolog, methyltransferase; coenzyme Q3 homolog, methyltransferase (yeast); coenzyme Q3 methyltransferase; coenzyme Q3, methyltransferase; Coq3; DHHB methyltransferase; DHHBMT; DHHB-MT; DHHBMTASE; DHHB-MTase; dihydroxyhexaprenylbenzoate methyltransferase; hexaprenyldihydroxybenzoate methyltransferase, mitochondrial; methyltransferase COQ3; polyprenyldihydroxybenzoate methyltransferase; Ubiquinone biosynthesis O-methyltransferase, mitochondrial; UG0215E05
Gene Symbols COQ3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COQ3. Recombinant protein control fragment (Product #RP-94830).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.