Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COQ4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560171 Supplier Invitrogen™ Supplier No. PA560171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERPRISTSTL DLGKLQSLPE GSLGREYLRF LDVNRVSPDT RAPTRFVDDE ELAYVIQRYR EVHDMLHTLL GMPTNILGEI VVKW Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

Component of the coenzyme Q biosynthetic pathway. May play a role in organizing a multi-subunit COQ enzyme complex required for coenzyme Q biosynthesis. Required for steady-state levels of other COQ polypeptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COQ4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Coq4
Gene Accession No. Q9Y3A0
Gene Alias A330041N06; CGI-92; Coenzyme Q biosynthesis protein 4 homolog; coenzyme Q biosynthesis protein 4 homolog {ECO:0000255; coenzyme Q4; coenzyme Q4 homolog; coenzyme Q4 homolog (yeast); COQ10D7; COQ4; D2Ertd97e; EST-MNCb4625; HAMAP-Rule:MF_03111}; ubiquinone biosynthesis protein COQ4 homolog, mitochondrial; ubiquinone biosynthesis protein COQ4 homolog, mitochondrial {ECO:0000255
Gene Symbols Coq4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COQ4. Recombinant protein control fragment (Product #RP-93039).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51117
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.