Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COQ5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561988 Supplier Invitrogen™ Supplier No. PA561988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGGRFLCLEF SQVNNPLISR LYDLYSFQVI PVLGEVIAGD WKSYQYLVES IRRFPSQEEF KDMIEDAGFH KVTYESLTSG IVAIHSGFKL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 94%.

Methyltransferase required for the conversion of 2-polyprenyl-6-methoxy-1,4-benzoquinol (DDMQH2) to 2-polyprenyl-3-methyl-6-methoxy-1,4-benzoquinol (DMQH2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COQ5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COQ5
Gene Accession No. Q5HYK3
Gene Alias 1810014G04Rik; 2-methoxy-6-polyprenyl-1,4-benzoquinol methylase, mitochondrial; coenzyme Q5 homolog, methyltransferase; coenzyme Q5 homolog, methyltransferase (S. cerevisiae); coenzyme Q5 homolog, methyltransferase (yeast); coenzyme Q5, methyltransferase; Coq5; D5Ertd33e; RGD1310857; Ubiquinone biosynthesis methyltransferase COQ5; ubiquinone biosynthesis methyltransferase COQ5, mitochondrial; Unknown (protein for MGC:142779)
Gene Symbols COQ5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COQ5. Recombinant protein control fragment (Product #RP-97681).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84274
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.