Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COQ6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100233 Supplier Invitrogen™ Supplier No. PA5100233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total COQ6.

COQ6 (Coenzyme Q10 monooxygenase 6) is also named as CGI-10 and belongs to the ubiH/COQ6 family. It is an evolutionarily conserved flavin-dependent monooxygenase required for biosynthesis of coenzyme Q10, or ubiquinone. It has 3 isoforms with the molecular mass of 51 kDa, 43 kDa and 49 kDa. In inner ear, Coq6 is expressed in spiral ganglion, stria vascularis cells, and spiral ligament. Defects in COQ6 are the cause of coenzyme Q10 deficiency, primary, type 6 (COQ10D6).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COQ6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Coq6
Gene Accession No. Q68FU7, Q8R1S0, Q9Y2Z9
Gene Alias 5930427M12Rik; AW742344; CGI10; CGI-10; Coenzyme Q10 monooxygenase 6; coenzyme Q6 homolog (yeast); coenzyme Q6 homolog, monooxygenase; coenzyme Q6 monooxygenase; coenzyme Q6, monooxygenase; COQ10D6; Coq6; ubiquinone biosynthesis monooxygenase COQ6; ubiquinone biosynthesis monooxygenase COQ6, mitochondrial
Gene Symbols Coq6
Host Species Rabbit
Immunogen A synthesized peptide derived from human COQ6(Accession Q9Y2Z9), corresponding to amino acid residues L54-W104.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217707, 299195, 51004
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.