Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COQ7 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299953R
Description
COQ7 Polyclonal specifically detects COQ7 in Human samples. It is validated for ELISA.Specifications
COQ7 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 550 | |
CAT5, CLK1, CLK-1, Coenzyme Q biosynthesis protein 7 homolog, coenzyme Q, 7 (rat, yeast) homolog, coenzyme Q7 homolog, ubiquinone (yeast), COQ7 coenzyme Q, 7 homolog ubiquinone, placental protein KG-20, Timing protein clk-1 homolog, ubiquinone biosynthesis protein COQ7 homolog | |
Produced in rabbits immunized with purified, recombinant COQ7 (Q99807-1; Ser 37-Leu 217). | |
0.1 mL | |
Stem Cell Markers | |
10229 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction