Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COQ7 Monoclonal Antibody (06)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530267
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human COQ7.
The protein encoded by this gene is similar to a mitochondrial di-iron containing hydroxylase in Saccharomyces cerevisiae that is involved with ubiquinone biosynthesis. Mutations in the yeast gene lead to slower development and longer life span. Alternatively spliced transcript variants have been found for this gene.
Specifications
COQ7 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q99807 | |
COQ7 | |
Recombinant Human COQ7 Protein (Ser37-Leu217). | |
100 μL | |
Primary | |
Human | |
Antibody |
ELISA | |
06 | |
Unconjugated | |
COQ7 | |
5-demethoxyubiquinone hydroxylase; 5-demethoxyubiquinone hydroxylase, mitochondrial; CAT5; CLK1; CLK-1; Coenzyme q (ubiquinone) biosynthetic enzyme (DHPB methyltransferase) 7; coenzyme Q biosynthesis protein 7 homolog; coenzyme Q7 homolog, ubiquinone; coenzyme Q7 homolog, ubiquinone (yeast); coenzyme Q7, hydroxylase; COQ10D8; COQ7; COQ7 coenzyme Q, 7 homolog ubiquinone; demethyl-Q 7; DMQ hydroxylase; placental protein KG-20; timing protein clk-1 homolog; Ubiquinone biosynthesis monooxygenase COQ7; ubiquinone biosynthesis protein COQ7 homolog | |
Mouse | |
Protein A | |
RUO | |
10229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction